Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL1RAPL1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $468.50
Specifications
Antigen | IL1RAPL1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
IL1RAPL1 Polyclonal specifically detects IL1RAPL1 in Human samples. It is validated for Western Blot.Specifications
IL1RAPL1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
IL1R8IL1RAPL, IL-1R9, IL-1RAPL1, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1 | |
IL1RAPL1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9NZN1 | |
11141 | |
Mouse myeloma cell line NS0-derived recombinant human IL1RAPL1, Leu19-Thr357, Accession # Q9NZN1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human IL1RAPL1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human IL1RAcP is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title