Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human IL6R ELISA Kit (Colorimetric)

Click to view available options
Quantity:
96 Tests
Description
The Human IL6R ELISA Kit (Colorimetric) measures human IL6R in cell culture supernatant, serum, plasma (edta, heparin), amniotic fluid, urine.
Specifications
Specifications
Assay Range | 0.08 - 5.0ng/mL |
Assay Sensitivity | 0.01ng/mL |
Product Type | ELISA Kit |
Sample Type | Amniotic Fluid, Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine |
Specificity | The interference of circulating factors of the immune systeme was evaluated by spiking these proteins at physiologically relevant concentrations into a human IL-6R positive serum. There was no crossreactivity detected. |
Target Species | Human |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 3570 |
Gene Symbol | IL6R |
Kit Contents | Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sIL-6R, HRP-Conjugate anti-human sIL-6R monoclonal antibody, Human sIL-6R Standard lyophilized, 5ng/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction