Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human immunodeficiency virus type 1 p24 Mouse anti-Virus, Clone: HM030, Abnova™

Catalog No. 89033558 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-558 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-558 Supplier Abnova Corporation Supplier No. MAB13595
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human immunodeficiency virus type 1 (HIV-1) p24.

  • High specificity
  • High reactivity

Specifications

Antigen Human immunodeficiency virus type 1 p24
Applications ELISA, Immunofluorescence, Lateral Flow
Classification Monoclonal
Clone HM030
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human immunodeficiency virus type 1 (HIV-1) p24.
Dilution ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Host Species Mouse
Purification Method Protein A purification
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with p24 of human immunodeficiency virus type 1.
Target Species Virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.