Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Importin alpha 2/KPNA2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB62071
This item is not returnable.
View return policy
Description
Importin alpha 2/KPNA2 Monoclonal specifically detects Importin alpha 2/KPNA2 in Human samples. It is validated for Immunocytochemistry.Specifications
Importin alpha 2/KPNA2 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
P52292 | |
KPNA2 | |
E. coli-derived recombinant human Importin alpha 2/KPNA2 Ser2-Asn132 Accession # P52292 | |
100 μg | |
Primary | |
Detects human Importin alpha 2/KPNA2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) KPNA1, rhKPNA3, rhKPNA4, rhKPNA5, or rhKPNB1 is observed. | |
Human | |
Purified |
Immunocytochemistry | |
682208 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
importin alpha 1, importin-alpha-P1, IPOA1, karyopherin alpha 2 (RAG cohort 1, importin alpha 1), Karyopherin subunit alpha-2, KPNA2, Pendulin, QIP2, QIP2importin alpha 2, RAG cohort 1, RAG cohort protein 1, RCH1, RCH1importin subunit alpha-2, SRP1, SRP1alpha, SRP1-alpha | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3838 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction