Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Importin alpha 2/KPNA2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB62071SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB62071SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB62071SP Supplier R&D Systems Supplier No. MAB62071SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Importin alpha 2/KPNA2 Monoclonal specifically detects Importin alpha 2/KPNA2 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Importin alpha 2/KPNA2
Applications Immunocytochemistry
Classification Monoclonal
Clone 682208
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P52292
Gene Alias importin alpha 1, importin-alpha-P1, IPOA1, karyopherin alpha 2 (RAG cohort 1, importin alpha 1), Karyopherin subunit alpha-2, KPNA2, Pendulin, QIP2, QIP2importin alpha 2, RAG cohort 1, RAG cohort protein 1, RCH1, RCH1importin subunit alpha-2, SRP1, SRP1alpha, SRP1-alpha
Gene Symbols KPNA2
Host Species Mouse
Immunogen E. coli-derived recombinant human Importin alpha 2/KPNA2 Ser2-Asn132 Accession # P52292
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3838
Test Specificity Detects human Importin alpha 2/KPNA2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) KPNA1, rhKPNA3, rhKPNA4, rhKPNA5, or rhKPNB1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.