Learn More
Human Importin alpha 2/KPNA2 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Importin alpha 2/KPNA2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL |
| Gene Accession No. | P52292 |
| Gene Alias | importin alpha 1, importin-alpha-P1, IPOA1, karyopherin alpha 2 (RAG cohort 1, importin alpha 1), Karyopherin subunit alpha-2, KPNA2, Pendulin, QIP2, QIP2importin alpha 2, RAG cohort 1, RAG cohort protein 1, RCH1, RCH1importin subunit alpha-2, SRP1, SRP1alpha, SRP1-alpha |
| Gene Symbols | KPNA2 |
| Host Species | Sheep |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.