Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Importin alpha 3/KPNA4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8204
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8204 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8204 Supplier R&D Systems Supplier No. MAB8204
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Importin alpha 3/KPNA4 Monoclonal specifically detects Importin alpha 3/KPNA4 in Human samples. It is validated for Western Blot, Simple Western.

Order Info

The product is shipped at ambient temperature.

Specifications

Antigen Importin alpha 3/KPNA4
Applications Western Blot
Classification Monoclonal
Clone 843124
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00629
Gene Alias importin alpha 3, Importin alpha Q1, importin subunit alpha-4, importin-alpha-Q1, IPOA3, karyopherin alpha 4 (importin alpha 3), KPNA4, MGC12217, MGC26703, Qip1, QIP12, QIP1Karyopherin subunit alpha-4, SRP3
Gene Symbols KPNA4
Host Species Mouse
Immunogen E. coli-derived recombinant human Importin alpha 3/KPNA4 Met1-Asn104 Accession # O00629
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3840
Test Specificity Detects human Importin alpha 3/KPNA4 in ELISA and Western blot. In direct ELISA, no cross-reactivity to human KPNA3 was detected.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.