Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Indoleamine 2,3-dioxygenase/IDO Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. IC6030U100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6030U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC6030U100 Supplier R&D Systems Supplier No. IC6030U100UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Indoleamine 2,3-dioxygenase/IDO Monoclonal specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen Indoleamine 2,3-dioxygenase/IDO
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 700838
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P14902
Gene Alias 3dioxygenase, EC 1.13.11.52, IDO, IDO1, IDOIDO-1, INDO, INDOindole 2,3-dioxygenase, Indoleamine 2, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase
Gene Symbols IDO1
Host Species Mouse
Immunogen E. coli-derived recombinant human Indoleamine 2,3-dioxygenase/IDO Ala2-Gly403 Accession # P14902
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3620
Test Specificity Detects human Indoleamine 2,3-dioxygenase/IDO in direct ELISA.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG1
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.