Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Indoleamine 2,3-dioxygenase/IDO Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10498110
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10498110 100 μg
MAB104981SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10498110 Supplier R&D Systems Supplier No. MAB104981100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Indoleamine 2,3-dioxygenase/IDO Monoclonal specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for ELISA.

Specifications

Antigen Indoleamine 2,3-dioxygenase/IDO
Applications ELISA
Classification Monoclonal
Clone 700826
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. P14902
Gene Alias 3dioxygenase, EC 1.13.11.52, IDO, IDO1, IDOIDO-1, INDO, INDOindole 2,3-dioxygenase, Indoleamine 2, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase
Host Species Mouse
Immunogen E. coli-derived recombinant human Indoleamine 2,3-dioxygenase/IDO, Ala2-Gly403, Accession # P14902
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3620
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.