Learn More
BD Human Induced Pluripotent Stem Cell Analysis and Sorting Kit
Catalog No. bdb562626
Click to view available options
Quantity:
50 Tests
For sorting and analysis of induced human pluripotent stem cell (hiPSC) cultures and cells in the process of being reprogrammed
  • Species Reactivity: Hu
  • Size: 50 tests
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Includes CD13 PerCP-Cy™5.5, SSEA-4 Alexa Fluor™ 647, TRA-1-60 PE, isotype control Antibodies, BD™ compBead Plus compensation particles
Quantity 50 Tests
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.