Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ING1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5758
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5758 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5758 Supplier R&D Systems Supplier No. AF5758
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

ING1 Polyclonal specifically detects ING1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen ING1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Gene Accession No. Q9UK53
Gene Alias growth inhibitor ING1, growth inhibitory protein ING1, inhibitor of growth family, member 1, inhibitor of growth protein 1, p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a, tumor suppressor ING1
Gene Symbols ING1
Host Species Goat
Immunogen E. coli-derived recombinant human ING1 Met213-Ser327 Accession # Q9UK53
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3621
Test Specificity Detects human ING1 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) ING2 and less than 1% cross-reactivity with with rhING3, rhING4, and rhING5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.