Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human INHBA Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF10024SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF10024SP 25 μg
AF10024100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF10024SP Supplier R&D Systems Supplier No. AF10024SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Antibody

Inhibin beta A specifically detects Inhibin beta A in Human samples. It is validated for ELISA.

Specifications

Antigen Inhibin beta A
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias Activin Beta A, Activin beta-A chain, beta A (activin A, activin AB alpha polypeptide), EDF, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FRP, FSH-releasing protein, Inhibin Beta A, inhibin beta A chain, Inhibin Subunit Beta A, inhibin, beta A, Inhibin, beta-1
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human INHBA
Purification Method Antigen Affinity-purified
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 3624.0
Test Specificity Detects human Inhibin beta A chain in direct ELISAs. In sandwich immunoassays, this antibody detects human Inhibin A when paired with the Inhibin alpha chain antibody, Catalog No. MAB338.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.