Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Inhibin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB338SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB338SP 25 μg
MAB338100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB338SP Supplier R&D Systems Supplier No. MAB338SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Inhibin Monoclonal specifically detects Inhibin in Human samples. It is validated for ELISA.

Specifications

Antigen Inhibin
Applications ELISA
Classification Monoclonal
Clone 130408R
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P05111
Gene Alias INHA, INHB, Inhibin A
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Inhibin alpha chain, Accession No. P08476
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Test Specificity Detects human Inhibin alpha chain in direct ELISAs. In sandwich immunoassays, this antibody captures the human Inhibin alpha chain and detects either human Inhibin A or human Inhibin B based on the detection antibody.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.