Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human INSL6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4430
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4430 100 μg
MAB4430SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4430 Supplier R&D Systems Supplier No. MAB4430
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

INSL6 Monoclonal specifically detects INSL6 in Human samples. It is validated for Western Blot.

Specifications

Antigen INSL6
Applications Western Blot
Classification Monoclonal
Clone 421401
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y581
Gene Alias Insulin-Like 6, Insulin-like peptide 5, Insulin-Like Peptide 6, Insulin-Like Peptide INSL6, Relaxin/Insulin-Like Factor 1, RIF1
Gene Symbols INSL6
Host Species Rat
Immunogen E. coli-derived recombinant human INSL6 Arg21-Tyr213 Accession # Q9Y581
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11172
Test Specificity Detects human INSL6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) INSL3 and rhINSL4 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.