Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Insulin R/CD220 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1544U100UG
This item is not returnable.
View return policy
Description
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for Flow Cytometry.Specifications
Insulin R/CD220 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_001073285 | |
INSR | |
Mouse myeloma cell line NS0-derived recombinant human Insulin R/CD220 His28-Lys944 Accession # NP_001073285 | |
RUO | |
3643 | |
Human | |
IgG2b |
Flow Cytometry | |
243524 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, INSR, insulin receptor, InsulinR, IR | |
Mouse | |
100 μg | |
Primary | |
Detects human Insulin R/CD220 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction