Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Insulin R/CD220 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB15442T100UG
This item is not returnable.
View return policy
Description
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for Flow Cytometry.Specifications
Insulin R/CD220 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, INSR, insulin receptor, InsulinR, IR | |
Mouse myeloma cell line NS0-derived recombinant human Insulin R/CD220 His28-Lys956 Accession # P06213 | |
100 μg | |
3643 | |
Human | |
Supernatant |
Flow Cytometry | |
243522 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P06213 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Insulin R/CD220 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction