Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Insulin R/CD220 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB15442SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB15442SP 25 μg
MAB15442100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB15442SP Supplier R&D Systems Supplier No. MAB15442SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Insulin R/CD220
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 243522
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/5 x 10^6 cells, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P06213
Gene Alias CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, INSR, insulin receptor, InsulinR, IR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Insulin R/CD220 His28-Lys956 Accession # P06213
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 3643
Test Specificity Detects human Insulin R/CD220 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.