Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Insulysin/IDE Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2496
This item is not returnable.
View return policy
Description
Insulysin/IDE Monoclonal specifically detects Insulysin/IDE in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
Insulysin/IDE | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P14735 | |
IDE | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Insulysin/IDE Met42-Leu1019 Accession # P14735 | |
100 μg | |
Primary | |
Detects human Insulysin/IDE in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
334501 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Abeta-degrading protease, EC 3.4.24, EC 3.4.24.56, FLJ35968, IDE, INSDEGM, Insulin protease, insulinase, insulin-degrading enzyme, Insulysin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3416 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction