Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Integrin alpha 1/CD49a Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA56761U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA56761U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA56761U100 Supplier R&D Systems Supplier No. FAB56761U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Integrin alpha 1/CD49a Monoclonal specifically detects Integrin alpha 1/CD49a in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha 1/CD49a
Applications Flow Cytometry
Classification Monoclonal
Clone 1090A
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P56199
Gene Alias CD49 antigen-like family member A, CD49a, CD49a antigen, integrin, alpha 1, ITGA1, Laminin and collagen receptor, very late activation protein 1, VLA-1, VLA1integrin alpha-1
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO derived recombinant human Integrin alpha 1 beta 1 Met1-Pro1141 (Integrin alpha 1) & Gln21-Asp728 (Integrin beta 1) Accession # P56199
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3672
Test Specificity Detects Integrin alpha 1/CD49a in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.