Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Integrin alpha 1/CD49a Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5676
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5676 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5676 Supplier R&D Systems Supplier No. AF5676
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Integrin alpha 1/CD49a Polyclonal specifically detects Integrin alpha 1/CD49a in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen Integrin alpha 1/CD49a
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready
Gene Accession No. P56199
Gene Alias CD49 antigen-like family member A, CD49a, CD49a antigen, integrin, alpha 1, ITGA1, Laminin and collagen receptor, very late activation protein 1, VLA-1, VLA1integrin alpha-1
Gene Symbols ITGA1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 1/CD49a Phe29-Pro1141 Accession # P56199
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3672
Test Specificity Detects human Integrin alpha 1/CD49a in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Integrin alpha 2, rhIntegrin alpha 11, recombinant mouse (rm) Integrin alpha 2, and rmIntegrin alpha 11 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.