Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha 11 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4235SP
This item is not returnable.
View return policy
Description
Integrin alpha 11 Polyclonal specifically detects Integrin alpha 11 in Human samples. It is validated for Western Blot.Specifications
Integrin alpha 11 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HsT18964, integrin alpha-11, integrin, alpha 11, ITGA11 | |
Goat | |
Affinity Purified | |
RUO | |
22801 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
NP_001004439 | |
ITGA11 | |
Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 11 Phe23-Pro1141 (Leu524Arg) Accession # NP_001004439 | |
25 μg | |
Primary | |
Detects human Integrin alpha 11 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse Integrin alpha 11 is observed and less than 1% cross-reactivity with recombinant human (rh) Integrin alpha 1 and rhIntegrin alpha 2 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction