Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Integrin alpha 11 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4235
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4235 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4235 Supplier R&D Systems Supplier No. AF4235
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Integrin alpha 11 Polyclonal specifically detects Integrin alpha 11 in Human samples. It is validated for Western Blot.

Specifications

Antigen Integrin alpha 11
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. NP_001004439
Gene Alias HsT18964, integrin alpha-11, integrin, alpha 11, ITGA11
Gene Symbols ITGA11
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 11 Phe23-Pro1141 (Leu524Arg) Accession # NP_001004439
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22801
Test Specificity Detects human Integrin alpha 11 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse Integrin alpha 11 is observed and less than 1% cross-reactivity with recombinant human (rh) Integrin alpha 1 and rhIntegrin alpha 2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.