Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Integrin alpha 2/CD49b Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA12332T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA12332T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA12332T100 Supplier R&D Systems Supplier No. FAB12332T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha 2/CD49b
Applications Flow Cytometry
Classification Monoclonal
Clone 430903
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P17301
Gene Alias alpha-2 subunit, CD49b, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), ITGA2, VLA 2, VLA-2 alpha
Gene Symbols ITGA2
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 2/CD49b Tyr30-Thr1132 Accession # P17301
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3673
Test Specificity Detects human Integrin alpha 2/CD49b.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.