Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Integrin alpha 2b/CD41 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB76161R10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB76161R10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB76161R10 Supplier R&D Systems Supplier No. FAB76161R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha 2b/CD41
Applications Flow Cytometry
Classification Monoclonal
Clone 2530A
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias CD41, CD41 antigen, CD41BHPA3, GP2B, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, HPA3, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGA2b, ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived heterodimer of Human Integrin alpha 2b (Leu32-Arg993, Accession P08514) and Human Integrin beta 3 (Gly27-Asp718, Accession P05106)
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3674
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.