Learn More
Human Integrin alpha 2b/CD41 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Integrin alpha 2b/CD41 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 745201 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Alias | CD41, CD41 antigen, CD41BHPA3, GP2B, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, HPA3, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGA2b, ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK |
| Gene Symbols | ITGA2B |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.