Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha 4/CD49d Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$200.50 - $1973.00
Specifications
Antigen | Integrin alpha 4/CD49d |
---|---|
Clone | 7.2R |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunocytochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1354SP
![]() |
R&D Systems
MAB1354SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1354100
![]() |
R&D Systems
MAB1354100 |
100 μg |
Each for $538.00
|
|
|||||
MAB1354500
![]() |
R&D Systems
MAB1354500 |
500 μg |
Each for $1,973.00
|
|
|||||
Description
Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
Integrin alpha 4/CD49d | |
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Mouse | |
Human | |
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), ITGA4, MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4, VLA-4 subunit alpha | |
ITGA4 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
7.2R | |
Flow Cytometry, Immunocytochemistry, CyTOF | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
3676 | |
DX-3 human melanoma cell line | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Integrin alpha 4/CD49d. Recognizes the human Integrin alpha 4/CD49d subunit. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title