Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha 5/CD49e Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1864G100UG
This item is not returnable.
View return policy
Description
Integrin alpha 5/CD49e Monoclonal specifically detects Integrin alpha 5/CD49e in Human samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 5/CD49e | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P08648 | |
ITGA5 | |
Mouse myeloma cell line NS0-derived recombinant human Integrin alpha 5/CD49e Phe42-Tyr995 Accession # P08648 | |
100 μg | |
Primary | |
Detects human Integrin alpha 5/CD49e. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
238307 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5, very late activation protein 5, alpha subunit, VLA-5, VLA-5 alpha | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3678 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction