Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Integrin alpha 5/CD49e Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1864SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1864SP 25 μg
MAB1864 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1864SP Supplier R&D Systems Supplier No. MAB1864SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Integrin alpha 5/CD49e Monoclonal specifically detects Integrin alpha 5/CD49e in Human samples. It is validated for Flow Cytometry, Adhesion Activation, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen Integrin alpha 5/CD49e
Applications Flow Cytometry, Adhesion Activation, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 238307
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Adhesion Activation, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P08648
Gene Alias CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5, very late activation protein 5, alpha subunit, VLA-5, VLA-5 alpha
Gene Symbols ITGA5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Integrin alpha 5/CD49e Phe42-Tyr995 Accession # P08648
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3678
Test Specificity Detects human Integrin alpha 5/CD49e.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.