Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha 6 beta 1 Heterodimer Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB7809T100UG
This item is not returnable.
View return policy
Description
Integrin alpha 6 beta 1 Monoclonal specifically detects Integrin alpha 6 beta 1 in Human samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 6 beta 1 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD49f, ITGA6B, VLA-6 | |
Chinese Hamster Ovary cell line, CHO-derived Human Integrin alpha 6 beta 1 heterodimer, Phe24-Ser1012(Integrin alpha 6) and Gln21-Asp728 (Integrin beta1), Accession # NP_000201 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2548B | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_000201 | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3655 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction