Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha L/CD11a Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB35951SP
This item is not returnable.
View return policy
Description
Integrin alpha L/CD11a Monoclonal specifically detects Integrin alpha L/CD11a in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Integrin alpha L/CD11a | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
CD11 antigen-like family member A, CD11a, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), ITGAL, Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, Ly-15, Ly-21, lymphocyte function-associated antigen 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3683 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Flow Cytometry, CyTOF | |
CR38 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ITGAL | |
Fibronectin purified human monocytes | |
25 μg | |
Primary | |
Detects human Integrin alpha L/CD11a in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Integrin alpha 1, alpha 2, alpha 3, alpha 4, alpha 5, alpha 6, alpha 8, alpha 9, alpha 10, alpha 11, alpha M, alpha V, alpha X, rhIntegrin beta 3, rhIntegrin alpha 6(x1)+ beta 4, alpha 7(x2)+ beta 1, alpha D+ beta 2, alpha E+ beta 7, recombinant mouse Integrin alpha 2b or alpha L is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction