Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha L/CD11a Antibody, R&D Systems™

Mouse Monoclonal Antibody
$184.70 - $444.00
Specifications
Antigen | Integrin alpha L/CD11a |
---|---|
Clone | 345908 |
Dilution | Western Blot 2 μg/mL, Simple Western 50 μg/mL, Immunohistochemistry 5-15 μg/mL |
Applications | Western Blot, Simple Western, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB35952100
![]() |
R&D Systems
MAB35952100 |
100 μg |
Each for $444.00
|
|
|||||
MAB35952SP
![]() |
R&D Systems
MAB35952SP |
25 μg |
Each for $184.70
|
|
|||||
Description
Integrin alpha L/CD11a Monoclonal antibody specifically detects Integrin alpha L/CD11a in Human samples. It is validated for Western Blot, Simple Western, ImmunohistochemistrySpecifications
Integrin alpha L/CD11a | |
Western Blot 2 μg/mL, Simple Western 50 μg/mL, Immunohistochemistry 5-15 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
CD11 antigen-like family member A, CD11a, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), ITGAL, Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, Ly-15, Ly-21, lymphocyte function-associated antigen 1 | |
Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha L, Tyr26-Met1089 (Tyr660Ile), Accession # CAA68747 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
345908 | |
Western Blot, Simple Western, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
CAA68747 | |
3683 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title