Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha V beta 3 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB12192G100UG
This item is not returnable.
View return policy
Description
Integrin alpha V beta 3 Monoclonal specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Flow Cytometry.Specifications
Integrin alpha V beta 3 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 | |
Chinese Hamster Ovary cell line CHO-derived human Integrin alpha V beta 3., Human Integrin alpha V (Phe31-Val992) and Human Integrin beta 3 (Gly27-Asp718), Accession # NP_002201 and AAA52589 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2549B | |
Flow Cytometry 0.25-1 ug/106 cells | |
NP_002201 and AAA52589 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
3685 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction