Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Integrin alpha V beta 3 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3050N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3050N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3050N100 Supplier R&D Systems Supplier No. FAB3050N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin alpha V beta 3 Monoclonal specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha V beta 3
Applications Flow Cytometry
Classification Monoclonal
Clone 23C6
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias antigen identified by monoclonal L230, CD51, CD51 antigen, CD61, GP3A, GPIIIA, INGRB3, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), ITGAV, ITGB3, MSK8, Vitronectin Receptor alpha, Vitronectin receptor subunit alpha, VNRA, VNRADKFZp686A08142
Gene Symbols ITGAV
Host Species Mouse
Immunogen Human osteoclasts
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3685
Test Specificity Detects human Integrin alpha V beta 3.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.