Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha V beta 3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 13 publications
$227.50 - $584.50
Specifications
| Antigen | Integrin alpha V beta 3 |
|---|---|
| Clone | 23C6 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation, Adhesion Blockade, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Adhesion Blockade, CyTOF |
Description
Integrin alpha V beta 3 Monoclonal specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Adhesion Blockade, CyTOF-ready.Specifications
| Integrin alpha V beta 3 | |
| LYOPH | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Adhesion Blockade, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| antigen identified by monoclonal L230, CD51, CD51 antigen, CD61, GP3A, GPIIIA, INGRB3, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), ITGAV, ITGB3, MSK8, Vitronectin Receptor alpha, Vitronectin receptor subunit alpha, VNRA, VNRADKFZp686A08142 | |
| ITGAV | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 23C6 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation, Adhesion Blockade, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3685 | |
| Human osteoclasts | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Integrin alpha V beta 3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title