Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Integrin alpha V beta 5 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2528G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2528G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2528G100 Supplier R&D Systems Supplier No. FAB2528G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin alpha V beta 5 Monoclonal specifically detects Integrin alpha V beta 5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha V beta 5
Applications Flow Cytometry
Classification Monoclonal
Clone P5H9
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias CD51, integrin subunit alpha V, ITGAV, ITGB5, MSK8, Vitronectin Receptor alpha, VNRA, VTNR
Gene Symbols ITGAV
Host Species Mouse
Immunogen HT1080 human fibrosarcoma cell line
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3685
Test Specificity Detects human Integrin alpha V beta 5. Recognizes the human Integrin alpha V beta 5 heterodimer and does not recognize the alpha V subunit in association with any other beta subunits.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.