Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha V beta 5 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 12 publications
Supplier: R&D Systems MAB2528SP
This item is not returnable.
View return policy
Description
Integrin alpha V beta 5 Monoclonal specifically detects Integrin alpha V beta 5 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry, CyTOF-ready.Specifications
Integrin alpha V beta 5 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
CD51, integrin subunit alpha V, ITGAV, ITGB5, MSK8, Vitronectin Receptor alpha, VNRA, VTNR | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3685 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry, CyTOF | |
P5H9 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ITGAV | |
HT1080 human fibrosarcoma cell line | |
25 μg | |
Primary | |
Detects human Integrin alpha V beta 5. Recognizes the human Integrin alpha V beta 5 heterodimer and does not recognize the alpha V subunit in association with any other beta subunits. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction