Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Integrin beta 5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3824
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3824 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3824 Supplier R&D Systems Supplier No. AF3824
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 2 publications

Integrin beta 5 Polyclonal specifically detects Integrin beta 5 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen Integrin beta 5
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Simple Western 50 μg/mL, Flow Cytometry 2.5 μg/106 cells, Immunocytochemistry 5 to 15 μg/mL, CyTOF-ready
Gene Accession No. P18084
Gene Alias FLJ26658, integrin beta-5, integrin, beta 5, ITGB5
Gene Symbols ITGB5
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO cell-derived recombinant human Integrin beta 5 Gly24-Asn719 Accession # P18084
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3693
Test Specificity Detects human Integrin beta 5 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) Integrin beta 5 is observed, and less than 5% cross-reactivity with recombinant human (rh) Integrin beta 1, rhIntegrin beta 3, rhIntegrin beta 4, rhIntegrin beta 6 and rmIntegrin beta 7 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.