Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin beta 7 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF4669
This item is not returnable.
View return policy
Description
Integrin beta 7 Polyclonal specifically detects Integrin beta 7 in Human samples. It is validated for Western Blot.Specifications
Integrin beta 7 | |
Polyclonal | |
Biotin | |
P26010 | |
ITGB7 | |
Chinese hamster ovary cell line CHO-derived recombinant human Integrin beta 7 long isoform Glu20-His723 Accession # P26010 | |
50 μg | |
Primary | |
Detects human Integrin beta 7 in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse Integrin beta 7 is observed and less than 1% cross-reactivity with recombinant human (rh) Integrin beta 2, rhIntegrin beta 5, and rhIntegrin beta 6 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7, ITGB7 | |
Sheep | |
Affinity Purified | |
RUO | |
3695 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction