Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin beta 7 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4669P
This item is not returnable.
View return policy
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Human samples. It is validated for Flow Cytometry.Specifications
Integrin beta 7 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7, ITGB7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3695 | |
Human | |
Purified |
Flow Cytometry | |
473207 | |
Flow Cytometry 10 uL/10^6 cells | |
P26010 | |
ITGB7 | |
Chinese hamster ovary cell line CHO-derived recombinant human Integrin beta 7 long isoform Glu20-His723 Accession # P26010 | |
100 Tests | |
Primary | |
Detects human Integrin beta 7 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Integrin beta 1, beta 2, beta 3, beta 4, beta 5, beta 6, beta 8, recombinant mouse Integrin beta 4, or beta 7 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction