Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin beta 8 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4775S100UG
This item is not returnable.
View return policy
Description
Integrin beta 8 Monoclonal specifically detects Integrin beta 8 in Human samples. It is validated for Flow Cytometry.Specifications
Integrin beta 8 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P26012 | |
ITGB8 | |
Mouse myeloma cell line NS0-derived recombinant human Integrin beta 8 Glu43-Arg684 (predicted) Accession # P26012 | |
RUO | |
3696 | |
Human | |
IgG2b |
Flow Cytometry | |
416922 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
integrin beta-8, integrin, beta 8, ITGB8 | |
Mouse | |
100 μg | |
Primary | |
Detects human Integrin beta 8 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Integrins beta 1, beta 2, beta 3, beta 4, beta 5, beta 6, beta 7, or recombinant mouse Integrin beta 8 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction