Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Intelectin-1/Omentin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $410.00
Specifications
Antigen | Intelectin-1/Omentin |
---|---|
Clone | 450520 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB4254SP
![]() |
R&D Systems
MAB4254SP |
25 μg |
Each for $200.50
|
|
|||||
MAB4254100
![]() |
R&D Systems
MAB4254100 |
100 μg |
Each for $410.00
|
|
|||||
Description
Intelectin-1/Omentin Monoclonal specifically detects Intelectin-1/Omentin in Human samples. It is validated for ELISA.Specifications
Intelectin-1/Omentin | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Q8WWA0 | |
55600 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
450520 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Endothelial lectin HL-1, FLJ20022, Galactofuranose-binding lectin, hIntL, HL-1, intelectin 1 (galactofuranose binding), intelectin-1, Intelectin1, INTLHL1, ITLN1, Itlna, ITLNITLN-1, LFR, LFRIntestinal lactoferrin receptor, omentin, Omentin-1 | |
E. coli-derived recombinant human Intelectin-1/Omentin Thr19-Ser298 Accession # Q8WWA0 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Intelectin-1/Omentin in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title