Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Intelectin-1/Omentin ELISA Kit (Colorimetric)
SDP

Catalog No. NB016703 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016703 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016703 Supplier Novus Biologicals™ Supplier No. NBP281144
Add to Cart
Add to Cart

This kit recognizes Human Intelectin-1/Omentin in samples.

No significant cross-reactivity or interference between Human Intelectin-1/Omentin and analogues was observed.

Specifications

Assay Range 0.63 to 40 ng/mL
Assay Sensitivity 0.38 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 55600
Gene Symbol ITLN1
Host Species Human
Quantity 1 Kit
Research Discipline Cardiovascular Biology, metabolism
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Intelectin-1/Omentin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.