Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Intelectin-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8004
This item is not returnable.
View return policy
Description
Intelectin-2 Monoclonal specifically detects Intelectin-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Intelectin-2 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q8WWU7 | |
ITLN2 | |
Chinese hamster ovary cell line CHO-derived recombinant human Intelectin-2 Met1-Tyr324 Accession # Q8WWU7 | |
100 μg | |
Primary | |
Detects human Intelectin-2 in ELISA. In Direct ELISA, no cross-reactivity with recombinant human (rh) Intelectin-1, recombinant mouse (rm) Intelectin-1, or rmIntelectin-2 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
874029 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HL-2, intelectin 2, Intelectin2, Intelectin-b, Itln1b, ITLN2, Itlnb, UNQ2789/PRO7179 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
142683 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction