Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-13 Recombinant Protein

Catalog No. ENRIL135
Click to view available options
Quantity:
5 μg

Recombinant Protein

Human Interleukin 13 (IL-13) Recombinant Protein, RIL135, from Invitrogen.

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4. Purity: >98% pure by SDS-PAGE and HPLC analysis. Activity: One unit of activity is <1.0ng/mL, as determined in a proliferation assay of human TF-1 cells, corresponding to a specific activity of >1x10^6 units/mg. Use: IL-13 exerts activity in the range of 0.1 to 10ng/mL for most in vitro applications. Reconstitution and Dilution: Reconstitute in distilled water to a concentration of 100ug/mL. After reconstitution, dilute cytokine in a neutral buffer containing carrier protein such as 5-10% FBS or 1-4% high purity BSA (fatty acid free fraction V). Lyophilized product is stable until expiration if stored desiccated below 0°C. Cytokines diluted as indicated above should be divided into aliquots and stored at -70°C. Recombinant proteins should be stored at concentrations >500ng/mL to reduce loss of activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35225
For Use With (Application) Control, ELISA, Immunoprecipitation, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 3596
Name Human IL-13
Quantity 5 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias interleukin-13
Common Name Human IL-13
Gene Symbol IL13
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.