Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IRAK1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4048SP
This item is not returnable.
View return policy
Description
IRAK1 Polyclonal specifically detects IRAK1 in Human samples. It is validated for Western Blot.Specifications
IRAK1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.11, EC 2.7.11.1, Il1rak, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle, Plpk | |
Goat | |
Antigen Affinity-purified | |
RUO | |
3654 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL | |
AAH54000 | |
IRAK1 | |
E. coli-derived recombinant human IRAK1 Met1-Ser693 Accession # AAH54000 | |
25 μg | |
Primary | |
Detects human IRAK1 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) IRAK2 and rhIRAK4 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction