Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IRAK1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4048
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4048 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4048 Supplier R&D Systems Supplier No. AF4048
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IRAK1 Polyclonal specifically detects IRAK1 in Human samples. It is validated for Western Blot.

Specifications

Antigen IRAK1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 20 ug/mL
Gene Accession No. AAH54000
Gene Alias EC 2.7.11, EC 2.7.11.1, Il1rak, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle, Plpk
Gene Symbols IRAK1
Host Species Goat
Immunogen E. coli-derived recombinant human IRAK1 Met1-Ser693 Accession # AAH54000
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3654
Test Specificity Detects human IRAK1 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) IRAK2 and rhIRAK4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.