Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IRAK4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB39191SP
This item is not returnable.
View return policy
Description
IRAK4 Monoclonal specifically detects IRAK4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
IRAK4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64 | |
E. coli-derived recombinant human IRAK4, Met1-Ser460, Accession # Q9NWZ3 | |
25 μg | |
Primary | |
Detects human IRAK4 in direct ELISAs. | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
413613 | |
Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL, CyTOF-ready | |
Q9NWZ3 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
51135 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction