Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IRAK4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3919
This item is not returnable.
View return policy
Description
IRAK4 Polyclonal specifically detects IRAK4 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
IRAK4 | |
Polyclonal | |
Unconjugated | |
Q9NWZ3 | |
IRAK4 | |
E. coli-derived recombinant human IRAK4 Met1-Ser460 Accession # Q9NWZ3 | |
100 μg | |
Primary | |
Detects human IRAK4 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) IRAK1 and rhIRAK2 is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 5-15 ug/mL | |
EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64 | |
Goat | |
Affinity Purified | |
RUO | |
51135 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction