Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IRF1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4830SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4830SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4830SP Supplier R&D Systems Supplier No. MAB4830SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IRF1 Monoclonal specifically detects IRF1 in Human samples. It is validated for Western Blot.

Specifications

Antigen IRF1
Applications Western Blot
Classification Monoclonal
Clone 686703
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10914
Gene Alias interferon regulatory factor 1, MAR, MARIRF-1
Gene Symbols IRF1
Host Species Mouse
Immunogen E. coli-derived recombinant human IRF1 Thr147-Pro325 Accession # P10914
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3659
Test Specificity Detects human IRF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IRF2 (aa 151-349), rhIRF3 (aa 206-427), rhIRF4 (aa 130-451), rhIRF5 (aa 214-395), rhIRF6 (aa 176-366), rhIRF7 (aa 289-503), rhIRF8 (aa 255-426), rhIRF9 (aa 238-393), or recombinant mouse IRF1 (aa 147-329) is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.