Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IRF4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $444.00
Specifications
Antigen | IRF4 |
---|---|
Clone | 503210 |
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB55251SP
![]() |
R&D Systems
MAB55251SP |
25 μg |
Each for $200.50
|
|
|||||
MAB55251100
![]() |
R&D Systems
MAB55251100 |
100 μg |
Each for $444.00
|
|
|||||
Description
IRF4 Monoclonal antibody specifically detects IRF4 in Human samples. It is validated for Western BlotSpecifications
IRF4 | |
Western Blot 2 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
interferon regulatory factor 4, IRF-4, LSIRF, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, MUM1, NF-EM5 | |
E. coli-derived recombinant human IRF4, Glu130-Glu451, Accession # Q15306 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
503210 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q15306 | |
3662 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title